Learn More
Medchemexpress LLC BAM 15 | 210302-17-3 | 99.8% | 340.29 | 100 MG
SDP
Catalog No. 5000343843
Encompass_Preferred
BAM 15 is a mitochondrial protonophore uncoupler and an oxidative phosphorylation (OXPHOS) uncoupler, intended for research purposes only. It effectively increases O2 consumption without elevating Reactive Oxygen Species (ROS) and maintains high uncoupled respiration rates across various cell lines. This compound also induces mitochondrial swelling, indicative of its protonophore activity, and has shown protective effects in vivo against kidney injury.
  • Mitochondrial protonophore uncoupler
  • Oxidative phosphorylation (OXPHOS) uncoupler
  • Increases O2 consumption without increasing Reactive Oxygen Species (ROS)
  • Maintains high uncoupled respiration across various cell lines
  • Induces mitochondrial swelling
  • Treated cells show higher viability compared to FCCP-treated cells
  • Protects against kidney injury in vivo
Catalog No. 50-003-43843 Supplier Medchemexpress LLC Supplier No. HY110284100MG
BAM 15 is a mitochondrial protonophore uncoupler and an oxidative phosphorylation (OXPHOS) uncoupler, intended for research purposes only. It effectively increases O2 consumption without elevating Reactive Oxygen Species (ROS) and maintains high uncoupled respiration rates across various cell lines. This compound also induces mitochondrial swelling, indicative of its protonophore activity, and has shown protective effects in vivo against kidney injury.
  • Mitochondrial protonophore uncoupler
  • Oxidative phosphorylation (OXPHOS) uncoupler
  • Increases O2 consumption without increasing Reactive Oxygen Species (ROS)
  • Maintains high uncoupled respiration across various cell lines
  • Induces mitochondrial swelling
  • Treated cells show higher viability compared to FCCP-treated cells
  • Protects against kidney injury in vivo