Learn More
Medchemexpress LLC BAM 15 | 210302-17-3 | 99.8% | 340.29 | 250 MG
SDP
Catalog No. 5000347702
Encompass_Preferred
BAM 15 is a mitochondrial protonophore uncoupler and an oxidative phosphorylation (OXPHOS) uncoupler. It offers several benefits for research applications.
  • Increases O2 consumption without increasing reactive oxygen species.
  • Maintains uncoupled respiration at high rates across a broad concentration range.
  • Fully increases mitochondrial respiration in the presence of oligomycin.
  • Induces mitochondrial swelling.
  • Treated cells are more viable than FCCP-treated cells across a broad dosing range.
  • Protects from kidney injury in vivo, reducing tubular necrosis and immune cell infiltration.
Catalog No. 50-003-47702 Supplier Medchemexpress LLC Supplier No. 250MGHY110284
BAM 15 is a mitochondrial protonophore uncoupler and an oxidative phosphorylation (OXPHOS) uncoupler. It offers several benefits for research applications.
  • Increases O2 consumption without increasing reactive oxygen species.
  • Maintains uncoupled respiration at high rates across a broad concentration range.
  • Fully increases mitochondrial respiration in the presence of oligomycin.
  • Induces mitochondrial swelling.
  • Treated cells are more viable than FCCP-treated cells across a broad dosing range.
  • Protects from kidney injury in vivo, reducing tubular necrosis and immune cell infiltration.