Learn More
Cayman Chemical BAM-22P (human, mouse, rat, bovine) (trifluoroacetate salt), Purity: ≥98%, 5 mg
SDP
Catalog No. 502913269
Encompass_Preferred
An endogenous neuropeptide derived from proenkephalin A; expressed in the adrenal medulla and at lower concentrations throughout the brain; binds the µ-, κ-, and δ-opioid receptors (Kis = 0.9, 2.9, and 7.2 nM, respectively); intracerebroventricular administration induces analgesia in a tail-flick assay in mice (ED50 = 6.4 nM); decreases the frequency of tail flicking, as well as paw lifting and licking in a rat model of formalin-induced nocifensive behavior at 5 nM/animal
Catalog No. 50-291-3269 Supplier Cayman Chemical Supplier No. 367235 MG
An endogenous neuropeptide derived from proenkephalin A; expressed in the adrenal medulla and at lower concentrations throughout the brain; binds the µ-, κ-, and δ-opioid receptors (Kis = 0.9, 2.9, and 7.2 nM, respectively); intracerebroventricular administration induces analgesia in a tail-flick assay in mice (ED50 = 6.4 nM); decreases the frequency of tail flicking, as well as paw lifting and licking in a rat model of formalin-induced nocifensive behavior at 5 nM/animal