Learn More
Medchemexpress LLC BAM(8-22) | 412961-36-5 | 99.89% | 1971.20 | 1 MG
SDP
Catalog No. 5000423318
Encompass_Preferred
BAM(8-22) is a proteolytically cleaved product of proenkephalin A and functions as a sensory neuron-specific receptor (SNSR) agonist. It is a potent activator of Mas-related G-protein-coupled receptors (Mrgprs), including MrgprC11 and hMrgprX1. This compound induces scratching in mice in an Mrgpr-dependent manner and exhibits an analgesic effect.
  • Potent activator of Mrgprs
  • Induces scratching in mice
  • Exhibits an analgesic effect
  • Can inhibit the activation of microglia
  • Increases miR-184 levels in LPS-treated BV-2 cells
Catalog No. 50-004-23318 Supplier Medchemexpress LLC Supplier No. HYP12411MG
BAM(8-22) is a proteolytically cleaved product of proenkephalin A and functions as a sensory neuron-specific receptor (SNSR) agonist. It is a potent activator of Mas-related G-protein-coupled receptors (Mrgprs), including MrgprC11 and hMrgprX1. This compound induces scratching in mice in an Mrgpr-dependent manner and exhibits an analgesic effect.
  • Potent activator of Mrgprs
  • Induces scratching in mice
  • Exhibits an analgesic effect
  • Can inhibit the activation of microglia
  • Increases miR-184 levels in LPS-treated BV-2 cells