Learn More
Medchemexpress LLC BAM(8-22) | 412961-36-5 | 99.89% | 1971.20 | 5 MG
SDP
Catalog No. 5000423374
Encompass_Preferred
BAM(8-22), a proteolytically cleaved product of proenkephalin A and sensory neuron-specific receptor (SNSR) agonist, is a potent activator of Mas-related G-protein-coupled receptors (Mrgprs), specifically MrgprC11 and hMrgprX1. It induces scratching in mice in an Mrgpr-dependent manner. Additionally, BAM(8-22) has an analgesic effect and can inhibit the activation of microglia.
  • Potent activator of Mas-related G-protein-coupled receptors (Mrgprs).
  • Induces scratching in mice in an Mrgpr-dependent manner.
  • Has an analgesic effect.
  • Inhibits the activation of microglia.
  • Increases miR-184 level in LPS-treated BV-2 cells.
  • Reduces Iba-1 and inflammatory cytokines in LPS-treated BV-2 cells.
  • Diminishes NMDA-evoked nocifensive behaviors and hyperalgesia in rat pain models.
  • Reduces NMDA-evoked expression of FLI and NADPH reactivity in the spinal dorsal horn.
Catalog No. 50-004-23374 Supplier Medchemexpress LLC Supplier No. HYP12415MG
BAM(8-22), a proteolytically cleaved product of proenkephalin A and sensory neuron-specific receptor (SNSR) agonist, is a potent activator of Mas-related G-protein-coupled receptors (Mrgprs), specifically MrgprC11 and hMrgprX1. It induces scratching in mice in an Mrgpr-dependent manner. Additionally, BAM(8-22) has an analgesic effect and can inhibit the activation of microglia.
  • Potent activator of Mas-related G-protein-coupled receptors (Mrgprs).
  • Induces scratching in mice in an Mrgpr-dependent manner.
  • Has an analgesic effect.
  • Inhibits the activation of microglia.
  • Increases miR-184 level in LPS-treated BV-2 cells.
  • Reduces Iba-1 and inflammatory cytokines in LPS-treated BV-2 cells.
  • Diminishes NMDA-evoked nocifensive behaviors and hyperalgesia in rat pain models.
  • Reduces NMDA-evoked expression of FLI and NADPH reactivity in the spinal dorsal horn.