Learn More
Medchemexpress LLC BAM(8-22) | 412961-36-5 | 99.9% | 1971.20 | 10 MG
SDP
Catalog No. 5000422155
Encompass_Preferred
BAM(8-22) is a proteolytically cleaved product of proenkephalin A and a sensory neuron-specific receptor (SNSR) agonist. It is a potent activator of Mas-related G-protein-coupled receptors (Mrgprs), specifically MrgprC11 and hMrgprX1. This compound induces scratching in mice in an Mrgpr-dependent manner, exhibits an analgesic effect, and inhibits microglial activation.
  • Potent activator of MrgprC11 and hMrgprX1
  • Induces scratching in mice in an Mrgpr-dependent manner
  • Exhibits an analgesic effect
  • Inhibits microglial activation
Catalog No. 50-004-22155 Supplier Medchemexpress LLC Supplier No. HYP124110MG
BAM(8-22) is a proteolytically cleaved product of proenkephalin A and a sensory neuron-specific receptor (SNSR) agonist. It is a potent activator of Mas-related G-protein-coupled receptors (Mrgprs), specifically MrgprC11 and hMrgprX1. This compound induces scratching in mice in an Mrgpr-dependent manner, exhibits an analgesic effect, and inhibits microglial activation.
  • Potent activator of MrgprC11 and hMrgprX1
  • Induces scratching in mice in an Mrgpr-dependent manner
  • Exhibits an analgesic effect
  • Inhibits microglial activation