Learn More
Cayman Chemical BAM15, 210302-17-3, 10 mg
SDP
Supplier:  Cayman Chemical 1781110
Encompass_Preferred
A mitochondrial protonophore uncoupler that does not depolarize the plasma membrane and protects mice from acute renal ischemic-reperfusion injury
Catalog No. NC1278482