Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bamaquimast | 135779-82-7 | MFCD00904310 | 98.97% | 303.36 g/mol | C16H21N3O3 | 1 ML
SDP

Catalog No. 5000245417
Encompass_Preferred

Bamaquimast is a research reagent described as a potent antiasthmatic agent. Supplied as a ready-to-use 10 mM solution in DMSO for laboratory research, it is intended for experimental studies; consult safety documentation and handling instructions before use.

  • Ready-to-use 10 mM solution in DMSO.
  • Supplied as a 1 mL aliquot.
  • High purity (98.97%).
  • Molecular weight 303.36 g/mol; formula C16H21N3O3.
  • CAS number 135779-82-7.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-45417 Supplier Medchemexpress LLC Supplier No. HY10142710MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bamaquimast is a research reagent described as a potent antiasthmatic agent. Supplied as a ready-to-use 10 mM solution in DMSO for laboratory research, it is intended for experimental studies; consult safety documentation and handling instructions before use.

  • Ready-to-use 10 mM solution in DMSO.
  • Supplied as a 1 mL aliquot.
  • High purity (98.97%).
  • Molecular weight 303.36 g/mol; formula C16H21N3O3.
  • CAS number 135779-82-7.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.