Learn More
Apexbio Technology LLC BAMB-4 891025-25-5 5mg
SDP
Catalog No. 502240097
Encompass_Preferred
BAMB-4 (CAS 891025-25-5) is a cell-permeable inhibitor of inositol-1 4 5-trisphosphate 3-kinase A (ITPKA) exhibiting an IC50 of 20 M ITPKA regulates actin filament dynamics and inositol phosphate signaling pathways by binding to and bundling actin filaments BAMB-4 is efficiently taken up by H1299 cells and demonstrates high specificity displaying inhibitory activity with increased Km and reduced Vmax for both InsP3 and ATP indicative of a mixed-type inhibition mechanism Among 42 kinases tested BAMB-4 selectively inhibited ITPKA This compound serves as a valuable tool for studying ITPKA-mediated signaling in cellular models
This item is not returnable. View return policy
Catalog No. 50-224-0097 Supplier Apexbio Technology LLC Supplier No. B47155
BAMB-4 (CAS 891025-25-5) is a cell-permeable inhibitor of inositol-1 4 5-trisphosphate 3-kinase A (ITPKA) exhibiting an IC50 of 20 M ITPKA regulates actin filament dynamics and inositol phosphate signaling pathways by binding to and bundling actin filaments BAMB-4 is efficiently taken up by H1299 cells and demonstrates high specificity displaying inhibitory activity with increased Km and reduced Vmax for both InsP3 and ATP indicative of a mixed-type inhibition mechanism Among 42 kinases tested BAMB-4 selectively inhibited ITPKA This compound serves as a valuable tool for studying ITPKA-mediated signaling in cellular models