Learn More
Selleck Chemical LLC BAMB-4-E0334-25MG
SDP
Catalog No. 503138325
Encompass_Preferred
BAMB-4 (ITPKA-IN-C14) is a new type of membrane permeable ITPKA inhibitor and IC50 is 37 M in the Kinase ADP-Glo assay
Catalog No. 50-313-8325 Supplier Selleck Chemical LLC Supplier No. E033425MG
BAMB-4 (ITPKA-IN-C14) is a new type of membrane permeable ITPKA inhibitor and IC50 is 37 M in the Kinase ADP-Glo assay