Learn More
Ted Pella Inc Bamboo Splints, 15cm (6") long, 100/pk
SDP
Catalog No. NC9691390
Encompass
A neat way to hold tissue for mincing or to move and sort samples. May also be used for cleaning apertures or as an applicator stick. Pointed on one end.
Catalog No. NC9691390 Supplier Ted Pella Inc Supplier No. 116
A neat way to hold tissue for mincing or to move and sort samples. May also be used for cleaning apertures or as an applicator stick. Pointed on one end.