Learn More
DYNAMISM INC Bambu Lab PETG HF White - 1kg
SDP
Catalog No. NC3595025
Encompass
Key Features:Smooth, soft textureAMS compatibleIncludes reusable spool Please Note: Requires drying to achieve optimal print quality.
Catalog No. NC3595025 Supplier DYNAMISM INC Supplier No. 33100
Key Features:Smooth, soft textureAMS compatibleIncludes reusable spool Please Note: Requires drying to achieve optimal print quality.