Learn More
MATTERHACKERS INC Bambu Lab PPS-CF Advanced Filament - 1.75mm (0.75kg)
SDP
Catalog No. NC3900794
Encompass
Elevate your 3D printing projects with the Bambu Lab PPS-CF Filament, designed for advanced performance and reliability. Perfect for professionals seeking exceptional results, this filament offers:Exceptional Heat Resistance: Withstands high temperatures, making it ideal for demanding applications.Superior Flame Retardancy: Ensures safety and durability in various environments.Outstanding Mechanical Properties: Provides strength and resilience for robust prints.Chemical and Solvent Resistance: Maintains integrity against harsh chemicals.
Catalog No. NC3900794 Supplier MATTERHACKERS INC Supplier No. MFDCUGZU
Elevate your 3D printing projects with the Bambu Lab PPS-CF Filament, designed for advanced performance and reliability. Perfect for professionals seeking exceptional results, this filament offers:Exceptional Heat Resistance: Withstands high temperatures, making it ideal for demanding applications.Superior Flame Retardancy: Ensures safety and durability in various environments.Outstanding Mechanical Properties: Provides strength and resilience for robust prints.Chemical and Solvent Resistance: Maintains integrity against harsh chemicals.