Learn More
Medchemexpress LLC Bambuterol-d9 hydrochloride | 1794810-59-5 | 99.88% | 412.96 | 1 MG
SDP
Catalog No. 5000386516
Encompass_Preferred
Bambuterol-d9 (hydrochloride) is the deuterium labeled Bambuterol. Bambuterol ((±)-Bambuterol) hydrochloride is a long-acting beta-adrenoceptor agonist (LABA) used in the treatment of asthma, and it is also a prodrug of terbutaline.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-86516 Supplier Medchemexpress LLC Supplier No. HY17501S1MG
Bambuterol-d9 (hydrochloride) is the deuterium labeled Bambuterol. Bambuterol ((±)-Bambuterol) hydrochloride is a long-acting beta-adrenoceptor agonist (LABA) used in the treatment of asthma, and it is also a prodrug of terbutaline.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.