Learn More
Medchemexpress LLC Bambuterol-d9 hydrochloride | 1794810-59-5 | 99.9% | 412.96 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY17501S5MG
Encompass_Preferred
Bambuterol-d9 hydrochloride is a deuterium-labeled form of Bambuterol, a long-acting beta-adrenoceptor agonist (LABA) and prodrug of terbutaline, primarily used in the treatment of asthma. This compound is also valuable as a tracer and internal standard for quantitative analysis in various spectroscopic methods.
  • Can be used as a tracer.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-86367