Learn More
Medchemexpress LLC HY-17501A 10mg Medchemexpress, Bambuterol hydrochloride CAS:81732-46-9 Purity:>98%
SDP
Catalog No. 502081970
Encompass_Preferred
Medchemexpress, HY-17501A 10mg Bambuterol hydrochloride CAS:81732-46-9 Bambuterol hydrochloride ((±)-Bambuterol hydrochloride; KWD-2183 hydrochloride) is a long acting beta-adrenoceptor agonist (LABA) used in the treatment of asthma; it also is a prodrug of terbutaline. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1970 Supplier Medchemexpress LLC Supplier No. HY17501A10MG
Medchemexpress, HY-17501A 10mg Bambuterol hydrochloride CAS:81732-46-9 Bambuterol hydrochloride ((±)-Bambuterol hydrochloride; KWD-2183 hydrochloride) is a long acting beta-adrenoceptor agonist (LABA) used in the treatment of asthma; it also is a prodrug of terbutaline. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.