Learn More
Medchemexpress LLC Bambuterol hydrochloride | 81732-46-9 | 403.90 | 10MM/1ML
SDP
Catalog No. 5000386861
Encompass_Preferred
Bambuterol hydrochloride is a long-acting beta-adrenoceptor agonist (LABA) used in the treatment of asthma. It also serves as a proagent of terbutaline. While contraindicated in pregnancy and individuals with seriously impaired liver function, it can be used by people with renal impairment with dose adjustments.
  • Acts as a long-acting beta-adrenoceptor agonist
  • Effective in the treatment of asthma
  • Functions as a proagent of terbutaline
  • Suitable for use by individuals with renal impairment (with dose adjustments)
Catalog No. 50-003-86861 Supplier Medchemexpress LLC Supplier No. HY17501A10MM/1ML
Bambuterol hydrochloride is a long-acting beta-adrenoceptor agonist (LABA) used in the treatment of asthma. It also serves as a proagent of terbutaline. While contraindicated in pregnancy and individuals with seriously impaired liver function, it can be used by people with renal impairment with dose adjustments.
  • Acts as a long-acting beta-adrenoceptor agonist
  • Effective in the treatment of asthma
  • Functions as a proagent of terbutaline
  • Suitable for use by individuals with renal impairment (with dose adjustments)