Learn More
Medchemexpress LLC Bambuterol hydrochloride | 81732-46-9 | 403.90 | 5 MG
SDP
Catalog No. 5000386864
Encompass_Preferred
Bambuterol hydrochloride is a long-acting beta-adrenoceptor agonist (LABA) and a prodrug of terbutaline, utilized in the treatment of asthma. It targets the Adrenergic Receptor and is involved in GPCR/G Protein and Neuronal Signaling pathways.
  • Acts as a long-acting beta-adrenoceptor agonist (LABA)
  • Targets the adrenergic receptor
  • Functions as a prodrug of terbutaline
  • Used in the treatment of asthma
  • Storage conditions include 4°C sealed, away from moisture; in solvent, -80°C for 6 months or -20°C for 1 month
Catalog No. 50-003-86864 Supplier Medchemexpress LLC Supplier No. HY17501A5MG
Bambuterol hydrochloride is a long-acting beta-adrenoceptor agonist (LABA) and a prodrug of terbutaline, utilized in the treatment of asthma. It targets the Adrenergic Receptor and is involved in GPCR/G Protein and Neuronal Signaling pathways.
  • Acts as a long-acting beta-adrenoceptor agonist (LABA)
  • Targets the adrenergic receptor
  • Functions as a prodrug of terbutaline
  • Used in the treatment of asthma
  • Storage conditions include 4°C sealed, away from moisture; in solvent, -80°C for 6 months or -20°C for 1 month