Learn More
Medchemexpress LLC Bambuterol hydrochloride | 81732-46-9 | MFCD03427293 | 99.9% | 403.90 | 100 MG
SDP
Catalog No. 5000386795
Encompass_Preferred
Bambuterol hydrochloride, also known as (±)-Bambuterol hydrochloride, is a long-acting beta-adrenoceptor agonist (LABA). It is primarily utilized in the treatment of asthma and functions as a proagent of terbutaline. Intended for research use only, this product is contraindicated in pregnancy and for individuals with severely impaired liver function, requiring dose adjustments for renal impairment.
  • Used in the treatment of asthma.
  • Functions as a proagent of terbutaline.
  • Intended for research use only.
  • Purity of 99.9%.
  • Available in solid form.
Catalog No. 50-003-86795 Supplier Medchemexpress LLC Supplier No. HY17501A100MG
Bambuterol hydrochloride, also known as (±)-Bambuterol hydrochloride, is a long-acting beta-adrenoceptor agonist (LABA). It is primarily utilized in the treatment of asthma and functions as a proagent of terbutaline. Intended for research use only, this product is contraindicated in pregnancy and for individuals with severely impaired liver function, requiring dose adjustments for renal impairment.
  • Used in the treatment of asthma.
  • Functions as a proagent of terbutaline.
  • Intended for research use only.
  • Purity of 99.9%.
  • Available in solid form.