Learn More
Cayman Chemical Bambuterol (hydrochloride), Purity: ≥95%, CAS: 81732-46-9, 100 mg
SDP
Catalog No. 502966716
Encompass_Preferred
A prodrug form of terbutaline; an inhibitor of BChE (IC50 = 3.9 nM for the human enzyme); increases the latency to histamine-induced collapse in guinea pigs at 1 and 2 mg/kg; completely inhibits ovalbumin-induced decreases in dynamic compliance and increases in pulmonary resistance in ovalbumin-sensitized guinea pigs at 10 mg/kg
Catalog No. 50-296-6716 Supplier Cayman Chemical Supplier No. 17887100 MG
A prodrug form of terbutaline; an inhibitor of BChE (IC50 = 3.9 nM for the human enzyme); increases the latency to histamine-induced collapse in guinea pigs at 1 and 2 mg/kg; completely inhibits ovalbumin-induced decreases in dynamic compliance and increases in pulmonary resistance in ovalbumin-sensitized guinea pigs at 10 mg/kg