Learn More
Medchemexpress LLC Bamethan hemisulfate | 5716-20-1 | 99.5% | 307.36 g/mol | C12H21NO6S | 10 MG
SDP
Catalog No. 5000298106
Encompass_Preferred
Bamethan hemisulfate is the hemisulfate salt of DL-bamethane, a small-molecule vasodilator supplied for research use. It is intended for studies of vascular function and blood flow and is provided as a well-characterized solid for analytical and experimental applications.
  • Vasodilator for cardiovascular and hypertension research.
  • High purity by HPLC suitable for analytical use.
  • Solid form for accurate weighing and formulation.
  • Stable when stored dry at recommended temperatures.
  • Characterized by CAS 5716-20-1 and molecular weight 307.36 g/mol.
Catalog No. 50-002-98106 Supplier Medchemexpress LLC Supplier No. HYB1538A10MG
Bamethan hemisulfate is the hemisulfate salt of DL-bamethane, a small-molecule vasodilator supplied for research use. It is intended for studies of vascular function and blood flow and is provided as a well-characterized solid for analytical and experimental applications.
  • Vasodilator for cardiovascular and hypertension research.
  • High purity by HPLC suitable for analytical use.
  • Solid form for accurate weighing and formulation.
  • Stable when stored dry at recommended temperatures.
  • Characterized by CAS 5716-20-1 and molecular weight 307.36 g/mol.