Learn More
New England Biolabs, Inc. BamHI Methyltransferase – 100 units
SDP
Catalog No. 50811644
Encompass
BamHI Methyltransferase modifies the internal cytosine residue (N4) of the sequence GGATCC. This enzyme comes from an E. coli strain that carries the cloned BamHI modification gene from Bacillus amyloliquefaciens H (ATCC 49763).
Catalog No. 50-811-644 Supplier New England Biolabs, Inc. Supplier No. M0223S
BamHI Methyltransferase modifies the internal cytosine residue (N4) of the sequence GGATCC. This enzyme comes from an E. coli strain that carries the cloned BamHI modification gene from Bacillus amyloliquefaciens H (ATCC 49763).