Learn More
New England Biolabs, Inc. BamHI – 10000 units
SDP
Supplier:  New England Biolabs, Inc. R0136S
Encompass
A restriction endonuclease that recognizes the sequence G^GATC_C.
Catalog No. 50-811-996