Learn More
New England Biolabs, Inc. BamHI – 10000 units
SDP
Catalog No. 50811997
Encompass
A restriction endonuclease that recognizes the sequence G^GATC_C.
Catalog No. 50-811-997 Supplier New England Biolabs, Inc. Supplier No. R0136T
A restriction endonuclease that recognizes the sequence G^GATC_C.