Learn More
New England Biolabs, Inc. BamHI – 50000 units
SDP
Catalog No. 50811995
Encompass
A restriction endonuclease that recognizes the sequence G^GATC_C.
Catalog No. 50-811-995 Supplier New England Biolabs, Inc. Supplier No. R0136M
A restriction endonuclease that recognizes the sequence G^GATC_C.