Learn More
New England Biolabs, Inc. BamHI-HF – 10000 units
SDP
Supplier:  New England Biolabs, Inc. R3136T
Encompass
A high fidelity restriction endonuclease that recognizes the sequence G^GATC_C. High Fidelity (HF) Restriction Enzymes have 100% activity in rCutSmart Buffer; single-buffer simplicity means more straightforward and streamlined sample processing. HF enzymes also exhibit dramatically reduced star activity. HF enzymes are all Time-Saver qualified and can therefore cut substrate DNA in 5-15 with the flexibility to digest overnight without degradation to DNA. Engineered with performance in mind, HF restriction enzymes are fully active under a broader range of conditions, minimizing off-target products, while offering flexibility in experimental design. NEB extensively performs quality controls on all standard and high-fidelity (HF) restriction enzymes.
Catalog No. NC0023817