Learn More
Molecular Research Center BAN 100 ML
SDP
Catalog No. NC9734505
Encompass
BAN is molecular biology grade 4-bromoanisole for phase separation in the TRI Reagent RT and RNAzol RT procedures.
Catalog No. NC9734505 Supplier Molecular Research Center Supplier No. BN 191
BAN is molecular biology grade 4-bromoanisole for phase separation in the TRI Reagent RT and RNAzol RT procedures.