Learn More
Med Vet International Merck Banamine Injection, 50mg/mL, 250mL
Catalog No. 502828056
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Banamine Solution (Flunixin Meglumine) is a potent non-narcotic, nonsteroidal, analgesic agent with anti-inflammatory and antipyretic activity. For intravenous or intramuscular use in horses and for intravenous use in beef and dairy cattle. Not for use in dry dairy cows and veal calves.
Catalog No. 50-282-8056 Supplier Med Vet International Supplier No. RXBANA250ML
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Banamine Solution (Flunixin Meglumine) is a potent non-narcotic, nonsteroidal, analgesic agent with anti-inflammatory and antipyretic activity. For intravenous or intramuscular use in horses and for intravenous use in beef and dairy cattle. Not for use in dry dairy cows and veal calves.