Learn More
Bioworld Banana AGS Basal Medium, 50 L
Catalog No. 50254828
Encompass_Preferred
Banana AGS Basal MediumContains Macronutrients, micronutrients, vitamins, and plant growth regulators required to culture bananasProvided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 4.71 g/LpH: 4.25–5.25Soluble in water
Catalog No. 50-254-828 Supplier Bioworld Supplier No. 306301344
Banana AGS Basal MediumContains Macronutrients, micronutrients, vitamins, and plant growth regulators required to culture bananasProvided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 4.71 g/LpH: 4.25–5.25Soluble in water