Learn More
Chemglass Life Sciences Banana Female to Male Extension Cable, 4ft
SDP
Catalog No. CG1872A21
Encompass_Preferred
Banana Female to Male Extension Cable, 4ft. Banana extension cable for CG-1872-A series of electrode adapters. Adapter permits CG-1872 series pH electrode(12mm OD) to be inserted either directly into smaller reactors or down through a CG-1872-B baffle.
Catalog No. CG1872A21 Supplier Chemglass Life Sciences Supplier No. CG1872A21
Banana Female to Male Extension Cable, 4ft. Banana extension cable for CG-1872-A series of electrode adapters. Adapter permits CG-1872 series pH electrode(12mm OD) to be inserted either directly into smaller reactors or down through a CG-1872-B baffle.