Learn More
Cole-Parmer Pomona Electronics 22497 3 ft (0.9 M) 50 Ohm Cable 0.195" (5mm) O.D. RG58C/U
Catalog No. 50993196 Shop All Cole-Parmer Products
Encompass
This BNC cable with molded strain relief is a 50 RG58C/U cable and ideal for most test and measurement applications. It is available for sale solely in the US and Canada.
Catalog No. 50-993-196 Supplier Cole-Parmer Supplier No. 2000746
This BNC cable with molded strain relief is a 50 RG58C/U cable and ideal for most test and measurement applications. It is available for sale solely in the US and Canada.