Learn More
Grainger BAND-IT Band Clamp Buckles, Clip-On, Stainless Steel, 1 in. Width, 25/Pk.
Catalog No. 502768362
Encompass_Preferred
These clip-on buckles with teeth are engineered for use with 1 in. wide banding material. Designed for secure fastening applications, these stainless steel buckles provide reliable performance in demanding industrial environments.
  • Manufactured from durable 201/301 stainless steel
  • Features a clip-on design with teeth for secure attachment
  • Compatible with 1 in. band width
  • Designed for use with band thickness of 0.044 in.
  • Suitable for applications with a maximum clamp diameter of 8 in.
  • Package quantity of 25
Catalog No. 50-276-8362 Supplier Grainger Supplier No. 16P353
These clip-on buckles with teeth are engineered for use with 1 in. wide banding material. Designed for secure fastening applications, these stainless steel buckles provide reliable performance in demanding industrial environments.
  • Manufactured from durable 201/301 stainless steel
  • Features a clip-on design with teeth for secure attachment
  • Compatible with 1 in. band width
  • Designed for use with band thickness of 0.044 in.
  • Suitable for applications with a maximum clamp diameter of 8 in.
  • Package quantity of 25