Learn More
Grainger Band Clamp Buckles 201/301SS 1 1/4 PK25
Catalog No. 502765533
Encompass_Preferred
The Band-It Band Clamp Buckles Clip-On are stainless steel band clamp buckles that provide a method of attachment for band clamps. They feature a clip-on design that allows for easy installation without the need for tools or fasteners. The buckles have a 304 stainless steel construction for durability and corrosion resistance. They accommodate band clamp widths up to 1/2" and have a temperature range of -65°F to +600°F. The buckles measure 5/8" wide x 7/8" long and have a weight capacity of 115 lbs. These clip-on buckles are suitable for creating an attachment point or joint using band clamps.
Catalog No. 50-276-5533 Supplier Grainger Supplier No. 16P354
The Band-It Band Clamp Buckles Clip-On are stainless steel band clamp buckles that provide a method of attachment for band clamps. They feature a clip-on design that allows for easy installation without the need for tools or fasteners. The buckles have a 304 stainless steel construction for durability and corrosion resistance. They accommodate band clamp widths up to 1/2" and have a temperature range of -65°F to +600°F. The buckles measure 5/8" wide x 7/8" long and have a weight capacity of 115 lbs. These clip-on buckles are suitable for creating an attachment point or joint using band clamps.