Learn More
Grainger BAND-IT C400 Ratchet Tensioner, Plated Steel, 1 ea
Catalog No. 502767463
Encompass_Preferred
This manual ratchet-style clamp tool is designed for tensioning and cutting stainless steel strapping. It operates without external power or attachments, making it a highly portable and reliable solution for securing straps on both flat and round surfaces. The tool is suitable for various applications, including sign hanging, insulation work, and cable or pipe banding.
  • Designed for stainless steel banding systems
  • Accommodates strapping widths from 0.1875 in. to 0.75 in.
  • Suitable for band thicknesses up to 0.030 in.
  • Constructed from durable plated steel
  • Manual tensioning mechanism ensures snug fit without overtightening
Catalog No. 50-276-7463 Supplier Grainger Supplier No. 16P345
This manual ratchet-style clamp tool is designed for tensioning and cutting stainless steel strapping. It operates without external power or attachments, making it a highly portable and reliable solution for securing straps on both flat and round surfaces. The tool is suitable for various applications, including sign hanging, insulation work, and cable or pipe banding.
  • Designed for stainless steel banding systems
  • Accommodates strapping widths from 0.1875 in. to 0.75 in.
  • Suitable for band thicknesses up to 0.030 in.
  • Constructed from durable plated steel
  • Manual tensioning mechanism ensures snug fit without overtightening