Learn More
Med Vet International 3M Coban 2 Layer Compression Bandage System, 2-9/10 Yard X 4 inch/4 inch X 5-1/10 Yard, Self-Adherent/Pull On Closure, Tan/White, Nonsterile, 35 To 40 mmhg, 2094N, 1/Box
Catalog No. 502830642
Encompass_Preferred
The Coban 2 bandage system is suitable for use on leg ulcers, wounds, edema and lymphedema It delivers consistent compression for up to 7 days to help medical conditions resolve fully Two-layer compression system remains securely in place to eliminate the need for adjustment It can be applied at full stretch to provide full coverage Comfort layer is 2-9/10 yards x 4 inches; compression layer is 4 inches x 5-1/10 yards First layer is white and second layer is tan Compression rating is 35 to 40 mmHg
Catalog No. 50-283-0642 Supplier Med Vet International Supplier No. 989726BX
The Coban 2 bandage system is suitable for use on leg ulcers, wounds, edema and lymphedema It delivers consistent compression for up to 7 days to help medical conditions resolve fully Two-layer compression system remains securely in place to eliminate the need for adjustment It can be applied at full stretch to provide full coverage Comfort layer is 2-9/10 yards x 4 inches; compression layer is 4 inches x 5-1/10 yards First layer is white and second layer is tan Compression rating is 35 to 40 mmHg