Learn More
Med Vet International Fluff Dressing Kerlix™ 4 X 4 Inch, Sterile, 12-Ply Square, 5072, 2/Pack, 25 Pack
Catalog No. 503044219
Encompass_Preferred
USP Type VII USP means gauze sponge meets United States Pharmacopeial Convention standards Twelve pairs Packed in sterile pairs 4 x 4 inches
Catalog No. 50-304-4219 Supplier Med Vet International Supplier No. 688663TR
USP Type VII USP means gauze sponge meets United States Pharmacopeial Convention standards Twelve pairs Packed in sterile pairs 4 x 4 inches