Learn More
Med Vet International Elastic Bandage 3M Ace 4 inch X 5.3 Foot Single Hook And Loop Closure Tan Nonsterile Standard Compression, 207604, 72/Case
Catalog No. 502831561
Encompass_Preferred
The tan 3M Ace elastic bandage provides support and stability for weak or injured joints such as ankles and wrists It's made of a stretchable and flexible material that's comfortable to wear for extended periods The 5? feet long bandage lays flat against the skin for a discreet fit that stays concealed beneath clothing This Ace elastic bandage with hook closure stays in place without clips The material is washable and re-usable 10.1 centimeters x 1.6 meters
Catalog No. 50-283-1561 Supplier Med Vet International Supplier No. 500545CS
The tan 3M Ace elastic bandage provides support and stability for weak or injured joints such as ankles and wrists It's made of a stretchable and flexible material that's comfortable to wear for extended periods The 5? feet long bandage lays flat against the skin for a discreet fit that stays concealed beneath clothing This Ace elastic bandage with hook closure stays in place without clips The material is washable and re-usable 10.1 centimeters x 1.6 meters