Learn More
Med Vet International Miltex Vantage Universal Bandage And Utility Scissors, 7.5in., Green Handle
Catalog No. 502829046
Encompass_Preferred
The Miltex Vantage V95-1029 universal bandage and utility scissors are 7.5" in length and have green handles. They are designed to cut bandage and gauze, cast material, metal sheets, and tough clothing material. Autoclavable to 143C or 290F for easy sterilization.
Catalog No. 50-282-9046 Supplier Med Vet International Supplier No. MILV951029
The Miltex Vantage V95-1029 universal bandage and utility scissors are 7.5" in length and have green handles. They are designed to cut bandage and gauze, cast material, metal sheets, and tough clothing material. Autoclavable to 143C or 290F for easy sterilization.