Learn More
Med Vet International Miltex Vantage Universal Bandage And Utility Scissors, 7.5in., Red Handle
Catalog No. 502829054
Encompass_Preferred
The Miltex Vantage V95-1025 universal bandage and utility scissors are 7.5" in length and have red handles. They are designed to cut bandage and gauze, cast material, metal sheets, and tough clothing material. Autoclavable to 143C or 290F for easy sterilization.
Catalog No. 50-282-9054 Supplier Med Vet International Supplier No. MILV951025
The Miltex Vantage V95-1025 universal bandage and utility scissors are 7.5" in length and have red handles. They are designed to cut bandage and gauze, cast material, metal sheets, and tough clothing material. Autoclavable to 143C or 290F for easy sterilization.