Learn More
Med Vet International Fluff Gauze Roll Sterilux AMD 4-1/2 Inch X 4.1 Yard 1 per Pack Sterile PHMB / Benzalkonium Chloride Roll Shape, 15713135, Each
Catalog No. 503044029
Encompass_Preferred
Bulky roll is 4½ x 147 ? inches and is packaged sterilely Antimicrobial: Bacteria colonization is prevented with the addition of PHMB (polyhexamethylene biguanide) and benzalkonium chloride
Catalog No. 50-304-4029 Supplier Med Vet International Supplier No. 1175431EA
Bulky roll is 4½ x 147 ? inches and is packaged sterilely Antimicrobial: Bacteria colonization is prevented with the addition of PHMB (polyhexamethylene biguanide) and benzalkonium chloride