Learn More
Med Vet International Cohesive Bandage 3M Coban 3 inch X 5 Yard Self-Adherent Closure Tan Nonsterile Standard Compression, 1583, Each
Catalog No. 502831045
Encompass_Preferred
3M's Coban cohesive bandage is a self-adherent wrap used to secure dressings and other devices; compress or protect wound sites; immobilize injuries, and more The self-adhering, elastic wrap remains in place so that it doesn't need to be readjusted This 3M cohesive wrap is lightweight and prevents drag on dressings, bandages and devices Its porous and breathable construction allows air to pass through to prevent the accumulation of heat This tan, 3-inch x 5-yard elastic cohesive bandage won't stick to skin, dressings or bandages It stretches to provide a snug fit and accommodate contours
Catalog No. 50-283-1045 Supplier Med Vet International Supplier No. 481237RL
3M's Coban cohesive bandage is a self-adherent wrap used to secure dressings and other devices; compress or protect wound sites; immobilize injuries, and more The self-adhering, elastic wrap remains in place so that it doesn't need to be readjusted This 3M cohesive wrap is lightweight and prevents drag on dressings, bandages and devices Its porous and breathable construction allows air to pass through to prevent the accumulation of heat This tan, 3-inch x 5-yard elastic cohesive bandage won't stick to skin, dressings or bandages It stretches to provide a snug fit and accommodate contours