Learn More
Med Vet International Cohesive Bandage 3M Coban Lf 2 inch X 5 Yard Self-Adherent Closure Tan Nonsterile Standard Compression, 2082, 36/Box
Catalog No. 502830820
Encompass_Preferred
3M's self-adhesive bandage wrap sticks to itself without clips, pins or additional adhesive, offering easy application You can tear the length needed by hand with no need for scissors These 2-inch x 5-yard bandages stay reliably in place to secure primary dressings and splints or to compress and immobilize injury sites The material is porous and lightweight for user comfort These Coban bandage wraps come in a tan color
Catalog No. 50-283-0820 Supplier Med Vet International Supplier No. 388423CS
3M's self-adhesive bandage wrap sticks to itself without clips, pins or additional adhesive, offering easy application You can tear the length needed by hand with no need for scissors These 2-inch x 5-yard bandages stay reliably in place to secure primary dressings and splints or to compress and immobilize injury sites The material is porous and lightweight for user comfort These Coban bandage wraps come in a tan color