Learn More
Med Vet International Cohesive Bandage 3M Coban Lf 4 inch X 5 Yard Self-Adherent Closure Tan Nonsterile Standard Compression, 2084, 18/Case
Catalog No. 502830941
Encompass_Preferred
The 3M™ self-adhesive bandage wraps stick to themselves without clips, pins or additional adhesive, offering easy application You can tear the length needed by hand with no need for scissors These 4-inch x 5-yard bandages stay reliably in place to secure primary dressings and splints or to compress and immobilize injury sites The material is porous, lightweight and latex-free for user comfort These Coban™ bandage wraps come in tan
Catalog No. 50-283-0941 Supplier Med Vet International Supplier No. 382060CS
The 3M™ self-adhesive bandage wraps stick to themselves without clips, pins or additional adhesive, offering easy application You can tear the length needed by hand with no need for scissors These 4-inch x 5-yard bandages stay reliably in place to secure primary dressings and splints or to compress and immobilize injury sites The material is porous, lightweight and latex-free for user comfort These Coban™ bandage wraps come in tan