Learn More
Med Vet International Cohesive Bandage 3M Coban Lf 6 inch X 5 Yard Self-Adherent Closure Tan Nonsterile Standard Compression, 2086, Each
Catalog No. 502832205
Encompass_Preferred
The self-adherent cohesive bandages stick to themselves without clips, pins or additional adhesive, offering easy application You can tear the length needed by hand with no need for scissors Elastic bandages stay reliably in place to secure primary dressings and splints or to compress and immobilize injury sites The material is porous and lightweight for user comfort 6 inches x 5 yards
Catalog No. 50-283-2205 Supplier Med Vet International Supplier No. 388427EA
The self-adherent cohesive bandages stick to themselves without clips, pins or additional adhesive, offering easy application You can tear the length needed by hand with no need for scissors Elastic bandages stay reliably in place to secure primary dressings and splints or to compress and immobilize injury sites The material is porous and lightweight for user comfort 6 inches x 5 yards