Learn More
Med Vet International Co-Flex Tape 2in X 5 yds., Each
Catalog No. 502522417
Encompass_Preferred
Assorted colors, adheres to itself requiring no clips or fasteners, will not stick to hair. Individually shrink wrapped. No choice of colors. All rolls are 5 yards in length. Contains Latex. Co-Flex is a registered trademark of Andover Coated Products. Can be used as vet wrap.
Catalog No. 50-252-2417 Supplier Med Vet International Supplier No. COFLEX2
Assorted colors, adheres to itself requiring no clips or fasteners, will not stick to hair. Individually shrink wrapped. No choice of colors. All rolls are 5 yards in length. Contains Latex. Co-Flex is a registered trademark of Andover Coated Products. Can be used as vet wrap.