Learn More
Med Vet International Elastic Bandage, 3-in x 5 yds Stretched Length
Catalog No. 502522397
Encompass_Preferred
Economical and reusable, elastic bandages offer balanced stretch and compression. Made of rubber-reinforced high quality cotton yarn, these bandages can be laundered and sterilized.
Catalog No. 50-252-2397 Supplier Med Vet International Supplier No. EB3
Economical and reusable, elastic bandages offer balanced stretch and compression. Made of rubber-reinforced high quality cotton yarn, these bandages can be laundered and sterilized.