Learn More
Med Vet International Elastic Bandage, 4-in x 5 yds Stretched Length
Supplier:  Med Vet International EB4
Encompass_Preferred
Economical and reusable, elastic bandages offer balanced stretch and compression. Made of rubber-reinforced high quality cotton yarn, these bandages can be laundered and sterilized.
Catalog No. 50-252-2395