Learn More
Med Vet International Fluff Bandage Roll Dukal™ 4-1/2 Inch X 4-1/10 Yard 1 per Pack Sterile 6-Ply Roll Shape, 645, Each
Catalog No. 503042912
Encompass_Preferred
The Dukal™ fluff bandage roll is made of 100% cotton, washed and fluff-dried for a soft texture The open crinkle weave wicks away moisture, allows airflow and increases absorbency Smooth, finished edges prevent loose threads and lint Sterile bandage roll conforms to body curves, adding bulk and cushioning to comfortably secure dressings 4-1/2 inches x 4-1/10 yards
Catalog No. 50-304-2912 Supplier Med Vet International Supplier No. 360488EA
The Dukal™ fluff bandage roll is made of 100% cotton, washed and fluff-dried for a soft texture The open crinkle weave wicks away moisture, allows airflow and increases absorbency Smooth, finished edges prevent loose threads and lint Sterile bandage roll conforms to body curves, adding bulk and cushioning to comfortably secure dressings 4-1/2 inches x 4-1/10 yards