Learn More
Med Vet International Fluff Dressing Kerlix™ 6 X 6-3/4 Inch, Sterile 12-Ply Rectangle, 3085-, 120/Case
Catalog No. 503042854
Encompass_Preferred
USP Type VII Pre-washed, fluff-dried woven gauze, with crinkle-weave pattern and diamond-fold construction offers loft, bulk and absorbency USP means gauze sponge meets United States Pharmacopeial Convention standards 6 x 6-3/4 inches
Catalog No. 50-304-2854 Supplier Med Vet International Supplier No. 401582CS
USP Type VII Pre-washed, fluff-dried woven gauze, with crinkle-weave pattern and diamond-fold construction offers loft, bulk and absorbency USP means gauze sponge meets United States Pharmacopeial Convention standards 6 x 6-3/4 inches