Learn More
Med Vet International Kerlix™ Sterile Fluff Bandage Roll, 6-Ply, 4-1/2 Inch x 4-1/10 Yard, 6715-, 100/Case
Catalog No. 503043388
Encompass_Preferred
Fluff Bandage Roll Kerlix™ 4-1/2 Inch X 4-1/10 Yard 1 per Pouch Sterile 6-Ply Roll Shape 6 ply Made of prewashed, fluff-dried 100% woven gauze with crinkle-weave pattern for loft and bulk to cushion and protect wound areas Finished edges reduce loose ends and lint reducing the risk of wound contamination and granulomas 4-1/2 inches x 4-1/10 yards
Catalog No. 50-304-3388 Supplier Med Vet International Supplier No. 10173CS
Fluff Bandage Roll Kerlix™ 4-1/2 Inch X 4-1/10 Yard 1 per Pouch Sterile 6-Ply Roll Shape 6 ply Made of prewashed, fluff-dried 100% woven gauze with crinkle-weave pattern for loft and bulk to cushion and protect wound areas Finished edges reduce loose ends and lint reducing the risk of wound contamination and granulomas 4-1/2 inches x 4-1/10 yards